siRNA Details
| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029010.1 | NP_083286.1 | 17 | 2037 | ||
| GenBank mRNA | AK029925.1 | - | 17 | 2037 | ||
| AK170689.1 | - | 1 | 1525 | |||
| BC021773.1 | - | 16 | 1790 | 4222994 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glb1l| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029010.1 | NP_083286.1 | 17 | 2037 | ||
| GenBank mRNA | AK029925.1 | - | 17 | 2037 | ||
| AK170689.1 | - | 1 | 1525 | |||
| BC021773.1 | - | 16 | 1790 | 4222994 | ||