siRNA Details
Gene Symbol: 4930426D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271580.1 | NP_001258509.1 | 4 | 1167 | ||
GenBank mRNA | AK015205.1 | - | 4 | 1168 | ||
AK156636.1 | - | 4 | 1199 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930426D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271580.1 | NP_001258509.1 | 4 | 1167 | ||
GenBank mRNA | AK015205.1 | - | 4 | 1168 | ||
AK156636.1 | - | 4 | 1199 |