siRNA Details
| Gene Symbol: Fam46c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142952.1 | NP_001136424.1 | 2 | 364 | ||
| GenBank mRNA | AK015259.1 | - | 1 | 168 | ||
| BC037072.1 | - | 2 | 354 | |||
| BC046309.1 | - | 2 | 358 | |||
| BC120717.1 | - | 2 | 355 | 40129640 | ||
| BC120719.1 | - | 2 | 355 | 40129642 | ||

Fam46c
Genomic Map