siRNA Details
| Gene Symbol: 4930435E12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029042.1 | NP_083318.1 | 2 | 928 | ||
| GenBank mRNA | AK015321.1 | - | 2 | 911 | ||
| AK077164.1 | - | 2 | 928 | |||
| BC115688.1 | - | 2 | 927 | 40104289 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930435E12Rik| Gene Symbol: 4930435E12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029042.1 | NP_083318.1 | 2 | 928 | ||
| GenBank mRNA | AK015321.1 | - | 2 | 911 | ||
| AK077164.1 | - | 2 | 928 | |||
| BC115688.1 | - | 2 | 927 | 40104289 | ||