siRNA Details
| Gene Symbol: 5830415F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029086.2 | NP_083362.1 | 2 | 230 | ||
| GenBank mRNA | AK017931.1 | - | 2 | 241 | ||
| AK044073.1 | - | 2 | 224 | |||
| AK052584.1 | - | 2 | 245 | |||
| AK153576.1 | - | 2 | 231 | |||
| BC092543.1 | - | 2 | 231 | 30315335 | ||

5830415F09Rik
Genomic Map