siRNA Details
Gene Symbol: Mdga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081160.1 | NP_001074629.1 | 16 | 5328 | ||
GenBank mRNA | BC056477.1 | - | 1 | 852 | ||
DQ788983.1 | - | 5757 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mdga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081160.1 | NP_001074629.1 | 16 | 5328 | ||
GenBank mRNA | BC056477.1 | - | 1 | 852 | ||
DQ788983.1 | - | 5757 |