siRNA Details
| Gene Symbol: Meikin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB987828.1 | - | 4 | 336 | ||
| AK015073.1 | - | 4 | 354 | |||
| BC107368.1 | - | 3 | 444 | 40056257 | ||
| BC107369.1 | - | 4 | 353 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Meikin| Gene Symbol: Meikin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB987828.1 | - | 4 | 336 | ||
| AK015073.1 | - | 4 | 354 | |||
| BC107368.1 | - | 3 | 444 | 40056257 | ||
| BC107369.1 | - | 4 | 353 | |||