siRNA Details
| Gene Symbol: Kbtbd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029116.2 | NP_083392.1 | 2 | 6690 | ||
| GenBank mRNA | AK129197.1 | - | 1 | 5654 | ||
| AK134761.1 | - | 1 | 2417 | |||
| BC038875.1 | - | 1 | 334 | |||
| BC049667.1 | - | 1 | 389 | |||
| BC080858.1 | - | 1 | 5661 | 6852896 | ||

Kbtbd11
Genomic Map