siRNA Details
| Gene Symbol: Kbtbd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029116.2 | NP_083392.1 | 2 | 5535 | ||
| GenBank mRNA | AK129197.1 | - | 1 | 4498 | ||
| AK134761.1 | - | 1 | 1262 | |||
| BC080858.1 | - | 1 | 4506 | 6852896 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kbtbd11| Gene Symbol: Kbtbd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029116.2 | NP_083392.1 | 2 | 5535 | ||
| GenBank mRNA | AK129197.1 | - | 1 | 4498 | ||
| AK134761.1 | - | 1 | 1262 | |||
| BC080858.1 | - | 1 | 4506 | 6852896 | ||