siRNA Details
| Gene Symbol: Cylc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162865.1 | NP_001156337.1 | 2 | 70 | ||
| GenBank mRNA | AK015651.1 | - | 2 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cylc2| Gene Symbol: Cylc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162865.1 | NP_001156337.1 | 2 | 70 | ||
| GenBank mRNA | AK015651.1 | - | 2 | 71 | ||