siRNA Details
| Gene Symbol: Lrriq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163559.1 | NP_001157031.1 | 3 | 257 | ||
| NM_029134.2 | NP_083410.1 | 3 | 257 | |||
| GenBank mRNA | AK015687.1 | - | 3 | 258 | ||
| AK158531.1 | - | 3 | 484 | |||
| BC051400.1 | - | 3 | 241 | 6310575 | ||

Lrriq1
Genomic Map