siRNA Details
Gene Symbol: Csmd2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029137.1 | NP_083413.1 | 5 | 544 | ||
GenBank mRNA | AK015502.1 | - | 5 | 545 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csmd2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029137.1 | NP_083413.1 | 5 | 544 | ||
GenBank mRNA | AK015502.1 | - | 5 | 545 |