siRNA Details
| Gene Symbol: 4930502E18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029142.1 | NP_083418.1 | 5 | 835 | ||
| GenBank mRNA | AK015678.1 | - | 5 | 841 | ||
| BC115663.1 | - | 5 | 835 | 40103303 | ||
| BC115664.1 | - | 5 | 835 | 40103304 | ||

4930502E18Rik
Genomic Map