siRNA Details
| Gene Symbol: 4930505A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100394.1 | NP_001093864.1 | 4 | 513 | ||
| GenBank mRNA | AK015698.1 | - | 5 | 581 | ||
| BC049719.1 | - | 4 | 492 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930505A04Rik| Gene Symbol: 4930505A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100394.1 | NP_001093864.1 | 4 | 513 | ||
| GenBank mRNA | AK015698.1 | - | 5 | 581 | ||
| BC049719.1 | - | 4 | 492 | |||