siRNA Details
| Gene Symbol: 4930513O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029174.1 | NP_083450.1 | 4 | 732 | ||
| GenBank mRNA | AK015788.1 | - | 4 | 733 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930513O06Rik| Gene Symbol: 4930513O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029174.1 | NP_083450.1 | 4 | 732 | ||
| GenBank mRNA | AK015788.1 | - | 4 | 733 | ||