siRNA Details
| Gene Symbol: Ralgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019250.2 | NP_062123.2 | 11, 12 | 1898 | ||
| XM_006233750.2 | XP_006233812.1 | 11, 12 | 2443 | |||
| XM_006233751.2 | XP_006233813.1 | 11, 12 | 4376 | |||
| XM_006233752.2 | XP_006233814.1 | 11, 12 | 1956 | |||
| XM_006233753.2 | XP_006233815.1 | 11, 12 | 1920 | |||
| XM_008761632.1 | XP_008759854.1 | 11, 12 | 2221 | |||
| GenBank mRNA | L07925.1 | - | 11, 12 | 1898 | ||

Ralgds
Genomic Map