siRNA Details
| Gene Symbol: Arhgap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081123.1 | NP_001074592.1 | 13 | 2959 | ||
| GenBank mRNA | AB073967.1 | - | 13 | 2826 | ||
| AK003157.1 | - | 1 | 57 | |||
| AK048167.1 | - | 13 | 2848 | |||
| BC090672.1 | - | 13 | 2930 | 30941825 | ||

Arhgap36
Genomic Map