siRNA Details
| Gene Symbol: Zfp820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029281.2 | NP_083557.2 | 5 | 1945 | ||
| GenBank mRNA | BC019219.1 | - | 5 | 1983 | 5066012 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp820| Gene Symbol: Zfp820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029281.2 | NP_083557.2 | 5 | 1945 | ||
| GenBank mRNA | BC019219.1 | - | 5 | 1983 | 5066012 | |