siRNA Details
| Gene Symbol: Fam183b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162878.1 | NP_001156350.1 | 1 | 112 | ||
| NM_029283.1 | NP_083559.1 | 1 | 112 | |||
| GenBank mRNA | AK013920.1 | - | 1 | 115 | ||
| AK157972.1 | - | 1 | 91 | |||
| BC048748.1 | - | 1 | 69 | |||
| BC061254.1 | - | 1 | 62 | |||
| BC094630.1 | - | 1 | 69 | |||
| BC117741.2 | - | 1 | 168 | |||
| BC126964.1 | - | 1 | 165 | |||
| FJ468008.1 | - | 1 | 51 | |||

Fam183b
Genomic Map