siRNA Details
| Gene Symbol: Syce3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162880.1 | NP_001156352.1 | 2 | 195 | ||
| NM_001162882.1 | NP_001156354.1 | 2 | 163 | |||
| GenBank mRNA | AK005702.1 | - | 2 | 117 | ||
| AK029781.1 | - | 2 | 164 | |||
| BC048615.1 | - | 2 | 195 | |||
| EU079025.1 | - | 2 | 156 | |||
| HQ130280.1 | - | 1 | 90 | |||

Syce3
Genomic Map