siRNA Details
| Gene Symbol: C1qbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019259.2 | NP_062132.2 | 6 | 798 | ||
| GenBank mRNA | AJ001102.1 | - | 6 | 839 | ||
| BC070510.1 | - | 6 | 798 | 7100505 | ||
| FQ212622.1 | - | 6 | 823 | |||
| FQ214537.1 | - | 6 | 848 | |||
| FQ219796.1 | - | 6 | 821 | |||
| FQ220621.1 | - | 6 | 841 | |||

C1qbp
Genomic Map