siRNA Details
| Gene Symbol: 1700009N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081095.1 | NP_001074564.1 | 2 | 1218 | ||
| GenBank mRNA | AK005799.1 | - | 2 | 1223 | ||
| BC050973.1 | - | 2 | 994 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700009N14Rik| Gene Symbol: 1700009N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081095.1 | NP_001074564.1 | 2 | 1218 | ||
| GenBank mRNA | AK005799.1 | - | 2 | 1223 | ||
| BC050973.1 | - | 2 | 994 | |||