siRNA Details
| Gene Symbol: Pou5f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029315.1 | NP_083591.1 | 1 | 1283 | ||
| GenBank mRNA | AK076753.1 | - | 1 | 1274 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou5f2| Gene Symbol: Pou5f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029315.1 | NP_083591.1 | 1 | 1283 | ||
| GenBank mRNA | AK076753.1 | - | 1 | 1274 | ||