siRNA Details
| Gene Symbol: 1700020N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029316.1 | NP_083592.1 | 2 | 278 | ||
| GenBank mRNA | AK006180.1 | - | 2 | 260 | ||
| BC049765.1 | - | 256 | ||||
| CA464409.1 | - | 2 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700020N15Rik| Gene Symbol: 1700020N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029316.1 | NP_083592.1 | 2 | 278 | ||
| GenBank mRNA | AK006180.1 | - | 2 | 260 | ||
| BC049765.1 | - | 256 | ||||
| CA464409.1 | - | 2 | 280 | |||