siRNA Details
| Gene Symbol: 1700020N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029316.1 | NP_083592.1 | 1 | 21 | ||
| GenBank mRNA | CA464409.1 | - | 1 | 23 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700020N15Rik| Gene Symbol: 1700020N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029316.1 | NP_083592.1 | 1 | 21 | ||
| GenBank mRNA | CA464409.1 | - | 1 | 23 | ||