siRNA Details
| Gene Symbol: Izumo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029317.1 | NP_083593.1 | 5 | 439 | ||
| GenBank mRNA | AK006268.1 | - | 5 | 413 | ||
| BC048526.1 | - | 6 | 476 | 6772588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Izumo2| Gene Symbol: Izumo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029317.1 | NP_083593.1 | 5 | 439 | ||
| GenBank mRNA | AK006268.1 | - | 5 | 413 | ||
| BC048526.1 | - | 6 | 476 | 6772588 | ||