siRNA Details
| Gene Symbol: 2310007L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029345.1 | NP_083621.1 | 6 | 775 | ||
| GenBank mRNA | AK009213.1 | - | 7 | 1098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310007L24Rik| Gene Symbol: 2310007L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029345.1 | NP_083621.1 | 6 | 775 | ||
| GenBank mRNA | AK009213.1 | - | 7 | 1098 | ||