siRNA Details
| Gene Symbol: 2310007L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029345.1 | NP_083621.1 | 6 | 616 | ||
| GenBank mRNA | AK009213.1 | - | 7 | 939 | ||
| AK153786.1 | - | 6 | 616 | |||
| BC116232.1 | - | 4 | 335 | |||
| BC116233.1 | - | 5 | 565 | |||

2310007L24Rik
Genomic Map