siRNA Details
Gene Symbol: Zbtb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029348.2 | NP_083624.2 | 4 | 7646 | ||
GenBank mRNA | AK147212.1 | - | 4 | 7600 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zbtb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029348.2 | NP_083624.2 | 4 | 7646 | ||
GenBank mRNA | AK147212.1 | - | 4 | 7600 |