siRNA Details
| Gene Symbol: Zbtb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029348.2 | NP_083624.2 | 4 | 4665 | ||
| GenBank mRNA | AK020335.1 | - | 1 | 842 | ||
| AK034066.1 | - | 1 | 641 | |||
| AK147212.1 | - | 4 | 4619 | |||
| AK173196.1 | - | 1 | 5001 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb4| Gene Symbol: Zbtb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029348.2 | NP_083624.2 | 4 | 4665 | ||
| GenBank mRNA | AK020335.1 | - | 1 | 842 | ||
| AK034066.1 | - | 1 | 641 | |||
| AK147212.1 | - | 4 | 4619 | |||
| AK173196.1 | - | 1 | 5001 | |||