siRNA Details
| Gene Symbol: Malsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029353.1 | NP_083629.1 | 3, 4 | 579 | ||
| GenBank mRNA | AK010366.1 | - | 3, 4 | 521 | ||
| AK031878.1 | - | 3, 4 | 530 | |||
| AK153503.1 | - | 2, 3 | 348 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Malsu1| Gene Symbol: Malsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029353.1 | NP_083629.1 | 3, 4 | 579 | ||
| GenBank mRNA | AK010366.1 | - | 3, 4 | 521 | ||
| AK031878.1 | - | 3, 4 | 530 | |||
| AK153503.1 | - | 2, 3 | 348 | |||