siRNA Details
| Gene Symbol: Wnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290311.1 | NP_001277240.1 | 6 | 1873 | ||
| NM_001290313.1 | NP_001277242.1 | 6 | 1873 | |||
| NM_029361.4 | NP_083637.2 | 6 | 1873 | |||
| GenBank mRNA | AK147550.1 | - | 6 | 1560 | ||

Wnk2
Genomic Map