siRNA Details
| Gene Symbol: 2010109A12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029363.1 | NP_083639.1 | 2, 3 | 299 | ||
| GenBank mRNA | AK008356.1 | - | 2, 3 | 303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010109A12Rik| Gene Symbol: 2010109A12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029363.1 | NP_083639.1 | 2, 3 | 299 | ||
| GenBank mRNA | AK008356.1 | - | 2, 3 | 303 | ||