siRNA Details
| Gene Symbol: Kxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029366.2 | NP_083642.1 | 3 | 278 | ||
| GenBank mRNA | AK013135.1 | - | 3 | 194 | ||
| AK178220.1 | - | 1 | 28 | |||
| BC038552.1 | - | 2 | 171 | |||
| BC048938.1 | - | 3 | 277 | 6399490 | ||
| BC053944.1 | - | 3 | 277 | |||
| Gene Symbol: Uba52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CK620747.1 | - | 177 | |||

Kxd1
Genomic Map