siRNA Details
| Gene Symbol: Tex30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029368.1 | NP_083644.1 | 7, 8 | 826 | ||
| GenBank mRNA | AK006487.1 | - | 5, 6 | 858 | ||
| AK160565.1 | - | 7, 8 | 827 | |||
| BC099557.1 | - | 6, 7 | 739 | 30916402 | ||
| BC116881.1 | - | 5, 6 | 1023 | 40126200 | ||
| BC119032.1 | - | 5, 6 | 1091 | 8733781 | ||

Tex30
Genomic Map