siRNA Details
Gene Symbol: Spata25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029370.1 | NP_083646.1 | 1 | 48 | ||
GenBank mRNA | AK006145.1 | - | 1 | 23 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spata25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029370.1 | NP_083646.1 | 1 | 48 | ||
GenBank mRNA | AK006145.1 | - | 1 | 23 |