siRNA Details
| Gene Symbol: 1700020A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163483.1 | NP_001156955.1 | 1 | 326 | ||
| NM_029375.1 | NP_083651.1 | 2 | 149 | |||
| GenBank mRNA | AK006141.1 | - | 1 | 131 | ||
| BC049710.1 | - | 1 | 326 | |||
| BC137914.1 | - | 1 | 322 | |||
| BC137915.1 | - | 1 | 322 | |||

1700020A23Rik
Genomic Map