siRNA Details
| Gene Symbol: 1700020A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163483.1 | NP_001156955.1 | 1 | 111 | ||
| GenBank mRNA | BC049710.1 | - | 1 | 111 | ||
| BC137914.1 | - | 1 | 107 | |||
| BC137915.1 | - | 1 | 107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700020A23Rik| Gene Symbol: 1700020A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163483.1 | NP_001156955.1 | 1 | 111 | ||
| GenBank mRNA | BC049710.1 | - | 1 | 111 | ||
| BC137914.1 | - | 1 | 107 | |||
| BC137915.1 | - | 1 | 107 | |||