siRNA Details
| Gene Symbol: Cldn22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029383.1 | NP_083659.1 | 1 | 146 | ||
| GenBank mRNA | AK008821.1 | - | 1 | 147 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn22| Gene Symbol: Cldn22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029383.1 | NP_083659.1 | 1 | 146 | ||
| GenBank mRNA | AK008821.1 | - | 1 | 147 | ||