siRNA Details
| Gene Symbol: Vwa5b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029401.1 | NP_083677.1 | 9 | 1296 | ||
| GenBank mRNA | AK016425.1 | - | 9 | 1317 | ||
| BC057376.1 | - | 8 | 1168 | |||
| BC132477.1 | - | 9 | 1296 | 40130754 | ||
| BC137851.1 | - | 9 | 1296 | 8860870 | ||
| BC144974.1 | - | 10 | 1351 | |||

Vwa5b1
Genomic Map