siRNA Details
Gene Symbol: Fam227a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029407.1 | NP_083683.1 | 18 | 3437 | ||
GenBank mRNA | AK135276.1 | - | 1 | 248 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam227a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029407.1 | NP_083683.1 | 18 | 3437 | ||
GenBank mRNA | AK135276.1 | - | 1 | 248 |