siRNA Details
| Gene Symbol: Fam227a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029407.1 | NP_083683.1 | 10 | 1299 | ||
| GenBank mRNA | AK017017.1 | - | 10 | 1138 | ||
| AK039547.1 | - | 9 | 1147 | |||
| AK039683.1 | - | 9 | 1147 | |||
| AK046478.1 | - | 8 | 1085 | |||
| BC118956.1 | - | 8 | 884 | |||

Fam227a
Genomic Map