siRNA Details
| Gene Symbol: Fam227a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029407.1 | NP_083683.1 | 12 | 1449 | ||
| GenBank mRNA | AK017017.1 | - | 12 | 1288 | ||
| AK046478.1 | - | 9 | 1152 | |||
| BC118956.1 | - | 9 | 951 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam227a| Gene Symbol: Fam227a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029407.1 | NP_083683.1 | 12 | 1449 | ||
| GenBank mRNA | AK017017.1 | - | 12 | 1288 | ||
| AK046478.1 | - | 9 | 1152 | |||
| BC118956.1 | - | 9 | 951 | |||