siRNA Details
| Gene Symbol: Rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080813.2 | NP_001074282.1 | 6 | 7049 | ||
| NM_029423.2 | NP_083699.2 | 5 | 5486 | |||
| GenBank mRNA | AK033627.1 | - | 1 | 1569 | ||
| AK131172.1 | - | 3 | 3992 | |||

Rab11fip1
Genomic Map