siRNA Details
| Gene Symbol: Tex26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029464.1 | NP_083740.1 | 7 | 987 | ||
| GenBank mRNA | AK016369.1 | - | 8 | 985 | ||
| AK087319.1 | - | 7 | 993 | |||
| AK161244.1 | - | 9 | 1172 | |||
| BC120819.1 | - | 7 | 987 | 40129742 | ||
| BC144997.1 | - | 7 | 1030 | |||
| BC144998.1 | - | 6 | 902 | |||
| BC144999.1 | - | 7 | 984 | 9053547 | ||

Tex26
Genomic Map