siRNA Details
| Gene Symbol: Tex26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029464.1 | NP_083740.1 | 5 | 602 | ||
| GenBank mRNA | AK087319.1 | - | 5 | 608 | ||
| AK161244.1 | - | 7 | 787 | |||
| BC120819.1 | - | 5 | 602 | 40129742 | ||
| BC144997.1 | - | 5 | 645 | |||
| BC144998.1 | - | 4 | 517 | |||
| BC144999.1 | - | 5 | 599 | 9053547 | ||

Tex26
Genomic Map