siRNA Details
Gene Symbol: 4930571K23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145759.1 | NP_001139231.1 | 1 | 1595 | ||
GenBank mRNA | AK016274.1 | - | 1 | 595 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930571K23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145759.1 | NP_001139231.1 | 1 | 1595 | ||
GenBank mRNA | AK016274.1 | - | 1 | 595 |