siRNA Details
| Gene Symbol: Adal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290811.1 | NP_001277740.1 | 10 | 1793 | ||
| NM_001290812.1 | NP_001277741.1 | 9 | 1687 | |||
| NM_029475.2 | NP_083751.1 | 10 | 1685 | |||
| GenBank mRNA | AK039677.1 | - | 9 | 1678 | ||
| AK049697.1 | - | 10 | 1786 | |||
| BC050879.1 | - | 10 | 1637 | 5687835 | ||
| BC052048.1 | - | 10 | 1637 | 5687835 | ||

Adal
Genomic Map