siRNA Details
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081428.1 | NP_001074897.1 | 18 | 3542 | ||
GenBank mRNA | AK014044.1 | - | 1 | 754 | ||
AK016375.1 | - | 10 | 1365 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081428.1 | NP_001074897.1 | 18 | 3542 | ||
GenBank mRNA | AK014044.1 | - | 1 | 754 | ||
AK016375.1 | - | 10 | 1365 |