siRNA Details
| Gene Symbol: Fam184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081428.1 | NP_001074897.1 | 12 | 2583 | ||
| GenBank mRNA | AK016375.1 | - | 3 | 313 | ||
| AK139673.1 | - | 7 | 1074 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam184a| Gene Symbol: Fam184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081428.1 | NP_001074897.1 | 12 | 2583 | ||
| GenBank mRNA | AK016375.1 | - | 3 | 313 | ||
| AK139673.1 | - | 7 | 1074 | |||